Calcium wolframate 99,9%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Calcium tungstate, Calcium tungsten oxide
IN1393-250G
IN1393-50G
APOSIN1393-250GEA
2130
DKK
APOSIN1393-250G
APOSIN1393-50G
Calcium wolframate 99,9%
Calcium wolframate
Formel:
CaO₄W Smeltepunkt: 1620 °C Massefylde: 6,06 g/cm³ (25 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00010913 CAS-nummer: 7790-75-2 EINECS: 232-219-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...