Bariumtitanium trioxid 99,9%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Barium titanate, Barium tantalum oxide, Barium titanate (IV), Barium titanium oxide

IN1226-100G IN1226-500G
APOSIN1226-100GEA 1620 DKK
APOSIN1226-100G APOSIN1226-500G
Bariumtitanium trioxid 99,9%
Bariumtitanium trioxid
Formel: BaO₃Ti
Molekylvægt: 233.21 g/mol
Smeltepunkt: 1620 °C
Massefylde: 6.08 g/cm³ (25 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003447
CAS-nummer: 12047-27-7
Merck Index: 12,01029

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR