K252c

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIS0505-1MG BIS0505-5MG
APOSBIS0505-1MGEA 1430 DKK
APOSBIS0505-1MG APOSBIS0505-5MG
K252c
K252c

Inhibits PKC with IC50 of 2.5 uM and PKA at about 10-fold higher concentrations, Bioorg. Med. Chem. Lett. 3: 1959-1964 (1993).

Formel: C₂₀H₁₃N₃O
Opbevaringstemperatur: Fryser
MDL Number: MFCD00236435
CAS-nummer: 85753-43-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR