4-Methylumbelliferyl-β-D-galactopyranoside

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 4-Methylumbelliferyl β-D-galactopyranoside, 4-Methyl-7-(((2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-2H-chromen-2-one

BIM1095-1G BIM1095-5G
APOSBIM1095-1GEA 240 DKK
APOSBIM1095-1G APOSBIM1095-5G
4-Methylumbelliferyl-β-D-galactopyranoside
4-Methylumbelliferyl-β-D-galactopyranoside

Fluorescent substrate for b-D-galactosidase.

Formel: C₁₆H₁₈O₈
Molekylvægt: 338.312 g/mol
Smeltepunkt: 260 °C (dec.)
Opbevaringstemperatur: Fryser
MDL Number: MFCD00036773
CAS-nummer: 6160-78-7
EINECS: 228-185-5

Bestil nu

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR