L-Lysine acetate salt 99,5%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

BIL0713-100G BIL0713-500G
APOSBIL0713-100GEA 327 DKK
APOSBIL0713-100G APOSBIL0713-500G
L-Lysine acetate salt 99,5%
L-Lysine acetate salt

USP, Pyrogen free

Formel: C₈H₁₈N₂O₄
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00039069
CAS-nummer: 57282-49-2
EINECS: 260-664-4

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR