Bolton-Hunter reagent (sulpho-SHPP)

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Sulfo-SHPP

BII101-1G BII101-10G
APOSBII101-1GEA 709 DKK
APOSBII101-1G APOSBII101-10G
Bolton-Hunter reagent (sulpho-SHPP)
Bolton-Hunter reagent (sulpho-SHPP)
Formel: C₁₃H₁₃NO₅
Smeltepunkt: 132…133 °C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00005515
CAS-nummer: 34071-95-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR