G418 disulfat solution 50 mg/ml
Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: G-418 sulphate, Geneticin®, G-418 disulphate, G418 sulphate
BIG0176-10ML BIG0176-50ML BIG0176-100ML
APOSBIG0176-10MLEA 818 DKK
APOSBIG0176-10ML APOSBIG0176-50ML APOSBIG0176-100ML
G418 disulfat solution 50 mg/ml
G418 disulfat
Formel: C₂₀H₄₄N₄O₁₈S₂
Molekylvægt: 692.72 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00058314
CAS-nummer: 108321-42-2