D-(-)-Lyxose

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

BIFS0109-1G BIFS0109-5G BIFS0109-10G
APOSBIFS0109-1GEA 218 DKK
APOSBIFS0109-1G APOSBIFS0109-5G APOSBIFS0109-10G
D-(-)-Lyxose
D-(-)-Lyxose
Formel: C₅H₁₀O₅
Molekylvægt: 150.13 g/mol
Smeltepunkt: 108…115 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00064362
CAS-nummer: 1114-34-7
EINECS: 214-212-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR