Swainsonine 99% min

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 8α,β-Octahydroindolizidine-1α,2α,8β-triol

BICL4200-1MG BICL4200-5MG
APOSBICL4200-1MGEA 1590 DKK
APOSBICL4200-1MG APOSBICL4200-5MG
Swainsonine 99% min
Swainsonine
Formel: C₈H₁₅NO₃
Opbevaringstemperatur: Fryser
CAS-nummer: 72741-87-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR