Kifunensine 99% min
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
BICL4125-10MG
BICL4125-5MG
BICL4125-25MG
APOSBICL4125-10MGEA
3880
DKK
APOSBICL4125-10MG
APOSBICL4125-5MG
APOSBICL4125-25MG
Kifunensine 99% min
Kifunensine
Formel:
C₈H₁₂N₂O₆ Opbevaringstemperatur: Fryser |
CAS-nummer:
109944-15-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...