Benzyl 2,3,4-Tri-O-benzyl-beta-D-glucopyranoside 98% min
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BICL4044-100MG
BICL4044-250MG
BICL4044-50MG
APOSBICL4044-100MGEA
3260
DKK
APOSBICL4044-100MG
APOSBICL4044-250MG
APOSBICL4044-50MG
Benzyl 2,3,4-Tri-O-benzyl-beta-D-glucopyranoside 98% min
Benzyl 2,3,4-Tri-O-benzyl-beta-D-glucopyranoside
Formel:
C₃₄H₃₆O₆ Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD03701135 CAS-nummer: 27851-29-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...