3-(Cyclohexylamino)-2-hydroxypropansulfonsyre (CAPSO) ≥97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 3-(Cyclohexylamino)-2-hydroxy-1-propanesulphonic acid

BIC1469-100G BIC1469-500G
APOSBIC1469-100GEA 654 DKK
APOSBIC1469-100G APOSBIC1469-500G
3-(Cyclohexylamino)-2-hydroxypropansulfonsyre (CAPSO) ≥97%
3-(Cyclohexylamino)-2-hydroxypropansulfonsyre (CAPSO)

Biological buffer - useful range 8.9-10.3


  • Forms a clear colourless solution in water (10%)


US Patent No. 4,246,194 (To Research Organics)

Formel: C₉H₁₉NO₄S
Smeltepunkt: 270…274 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00041778
CAS-nummer: 73463-39-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR