Coelenterazine h

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIC1028-1MG BIC1028-5MG
APOSBIC1028-1MGEA 0 DKK
APOSBIC1028-1MG APOSBIC1028-5MG
Coelenterazine h
Coelenterazine h
Formel: C₂₆H₂₁N₃O₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00798394
CAS-nummer: 50909-86-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR