5-Bromo-4-chloro-3-indolyl sulphate potassium salt
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
5-Bromo-4-chloro-1H-indol-3-yl sulphate·potassium salt
BIB6007-100MG
BIB6007-1G
BIB6007-500MG
APOSBIB6007-100MGEA
1480
DKK
APOSBIB6007-100MG
APOSBIB6007-1G
APOSBIB6007-500MG
5-Bromo-4-chloro-3-indolyl sulphate potassium salt
5-Bromo-4-chloro-3-indolyl sulphate potassium salt
Formel:
C₈H₄BrClKNO₄S Molekylvægt: 364.635 g/mol Opbevaringstemperatur: Fryser |
MDL Number:
MFCD00056929 CAS-nummer: 6578-07-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...