Alizaringul GG

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIA9252-10G BIA9252-25G BIA9252-5G
APOSBIA9252-10GEA 273 DKK
APOSBIA9252-10G APOSBIA9252-25G APOSBIA9252-5G
Alizaringul GG
Alizaringul GG

C.I. 14025

Formel: C₁₃H₈N₃NaO₅
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00044746
CAS-nummer: 584-42-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR