N-Acetyl-L-leucine
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
N-Ac-Leu-OH, Ac-Leu-OH
BIA1765-100G
BIA1765-250G
BIA1765-25G
APOSBIA1765-100GEA
567
DKK
APOSBIA1765-100G
APOSBIA1765-250G
APOSBIA1765-25G
N-Acetyl-L-leucine
N-Acetyl-L-leucine
Formel:
C₈H₁₅NO₃ Smeltepunkt: 187…190 °C Opbevaringstemperatur: Fryser |
MDL Number:
MFCD00065131 CAS-nummer: 1188-21-2 EINECS: 214-706-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...