1-Aminobenzotriazol

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

BIA0115-1G BIA0115-250MG
APOSBIA0115-1GEA 2460 DKK
APOSBIA0115-1G APOSBIA0115-250MG
1-Aminobenzotriazol
1-Aminobenzotriazole

Suicide inhibitor of cytochrome P450 and chloroperoxidase

Formel: C₆H₆N₄
Smeltepunkt: 81…85 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00132902
CAS-nummer: 1614-12-6

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR