Chromogenic media for colourful microbial detection, CHROMagar™
Leverandør: CHROMAGAR
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ST162 YE492 CP572 AQ056 BC732 CQ382 CQ392 EB042 EC168 EC169-25 EE222 EE223-25 EF322 EF323-25 EF342 EL382 LK970 LM852 MC122 MS25P MS25N PF652 PS832 RR702 RR703-25 SA162 SA163-25 SQ001 TX182 VB912 VB913-25 CP573-25 SU702 SU703-25
ST162EA 4880 DKK
ST162 YE492 CP572 AQ056 BC732 CQ382 CQ392 EB042 EC168 EC169-25 EE222 EE223-25 EF322 EF323-25 EF342 EL382 LK970 LM852 MC122 MS25P MS25N PF652 PS832 RR702 RR703-25 SA162 SA163-25 SQ001 TX182 VB912 VB913-25 CP573-25 CHRASU702 CHRASU703-25
Chromogenic media for colourful microbial detection, CHROMagar™
Mikrobiologiske medier Dehydreret Mikrobiologisk medie
Chromogenic technology applied to culture media allows you to differentiate and easily identify, inside one single plate, the growth of different microorganisms.
  • Flexibility of use: Prepare only the quantity you need
  • Very easy to prepare
  • Long shelf life: 18 months
  • Cost saving for overall analysis
  • No need for pH adjustment
  • Supplements always included in the pack
Dr A. Rambach, founder of CHROMagar™, developed the use of this technology in food microbiology. The main advantage of using chromogenic media is the ability to distinguish with the naked eye different microorganisms growing on the same plate.
All CHROMagar™ products are dehydrated culture media. The unit size of every packs is indicated in volume (litres or millilitres) that shows the volume of culture medium that you can prepare: With one bottle you can prepare 5 or 25 L of culture media.
After the preparation of the medium starting from the powder, the 5 L pack allows you to prepare around 250 plates, while the 25 L allows you to prepare 1250 plates.
Every chromogen exhibits its own distinctive colour; the result is a very specific and distinctive colouring of different microorganisms which become clearly distinguishable to the naked eye under normal lighting conditions.
The 'Rambach Agar', available in this product range, is still one of the most used and famous chromogenic culture media for detection and isolation of Salmonella spp.
Leveringsinformation: Supplements always included in the pack.
Emballering: 5 L pack = dehydrated culture media to prepare 5 L of agar or broth or 25 L pack = dehydrated culture media to prepare 25 L of agar or broth.