1H-Perfluorononane 98%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
PC9754-1G
PC9754-5G
PC9754-10G
APOSPC9754-1GEA
1920
DKK
APOSPC9754-1G
APOSPC9754-5G
APOSPC9754-10G
1H-Perfluorononane 98%
1H-Perfluorononane
Formel:
C₉HF₁₉ Kogepunkt: 138 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD08458171 CAS-nummer: 375-94-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...