2-[4-(4-Fluorobenzoyl)phenyl]-2-(4-methylphenyl)acetonitrile 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PC9323-1G PC9323-500MG PC9323-5G
APOSPC9323-1GEA 2270 DKK
APOSPC9323-1G APOSPC9323-500MG APOSPC9323-5G
2-[4-(4-Fluorobenzoyl)phenyl]-2-(4-methylphenyl)acetonitrile 97%
2-[4-(4-Fluorobenzoyl)phenyl]-2-(4-methylphenyl)acetonitrile
Formel: C₂₂H₁₆FNO
Smeltepunkt: 93…95 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00792166
CAS-nummer: 339115-18-3

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR