3-Fluoroprop-1-yl methanesulphonate 98%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
3-Fluoropropyl methanesulphonate, 3-Fluoroprop-1-yl mesylate, 3-Fluoroprop-1-yl methanesulphonate
PC9175-1G
PC9175-5G
PC9175-250MG
APOSPC9175-1GEA
763
DKK
APOSPC9175-1G
APOSPC9175-5G
APOSPC9175-250MG
3-Fluoroprop-1-yl methanesulphonate 98%
3-Fluoroprop-1-yl methanesulphonate
Formel:
C₄H₉FO₃S Kogepunkt: 116 °C (15 mm Hg) Flammepunkt: 109 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD01715281 CAS-nummer: 372-04-3 UN nummer: 2810 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...