7-(Trifluoromethyl)-1,2,3,4-tetrahydroisoquinoline 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PC911168-1G PC911168-250MG
APOSPC911168-1GEA 4260 DKK
APOSPC911168-1G APOSPC911168-250MG
7-(Trifluoromethyl)-1,2,3,4-tetrahydroisoquinoline 97%
7-(Trifluoromethyl)-1,2,3,4-tetrahydroisoquinoline
Formel: C₁₀H₁₀F₃N
Molekylvægt: 201.188 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD08059296
CAS-nummer: 199678-32-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR