2-(Chloromethyl)-3-fluoroanisole 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PC910910-1G PC910910-5G
APOSPC910910-1GEA 1640 DKK
APOSPC910910-1G APOSPC910910-5G
2-(Chloromethyl)-3-fluoroanisole 95%
2-(Chloromethyl)-3-fluoroanisole
Formel: C₈H₈ClFO
Molekylvægt: 174.6 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD13185598
CAS-nummer: 339586-05-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR