4-Chloro-2-methyl-5-(trifluoromethyl)aniline 95%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
PC910371-1G
PC910371-250MG
APOSPC910371-1GEA
3610
DKK
APOSPC910371-1G
APOSPC910371-250MG
4-Chloro-2-methyl-5-(trifluoromethyl)aniline 95%
4-Chloro-2-methyl-5-(trifluoromethyl)aniline
Formel:
C₈H₇ClF₃N Molekylvægt: 209.596 g/mol Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD09907679 CAS-nummer: 1379346-32-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...