4-Fluoro-3-pyrrolidinobenzaldehyde 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PC908773-1G PC908773-5G
APOSPC908773-1GEA 1050 DKK
APOSPC908773-1G APOSPC908773-5G
4-Fluoro-3-pyrrolidinobenzaldehyde 95%
4-Fluoro-3-pyrrolidinobenzaldehyde
Formel: C₁₁H₁₂FNO
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD13191603
CAS-nummer: 1197193-31-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR