4,5,6,7-Tetrahydro-3-(trifluoromethyl)-2H-indazole 95%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
PC906940-1G
PC906940-250MG
APOSPC906940-1GEA
3120
DKK
APOSPC906940-1G
APOSPC906940-250MG
4,5,6,7-Tetrahydro-3-(trifluoromethyl)-2H-indazole 95%
Formel:
C₈H₉F₃N₂ Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD06496299 CAS-nummer: 890005-22-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...