Potassium cyclopentoxymethyltrifluoroborate 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Potassium ((cyclopentyloxy)methyl)trifluoroborate

PC905473-1G PC905473-5G
APOSPC905473-1GEA 428 DKK
APOSPC905473-1G APOSPC905473-5G
Potassium cyclopentoxymethyltrifluoroborate 95%
Potassium cyclopentoxymethyltrifluoroborate
Formel: C₆H₁₁BF₃O·K
Molekylvægt: 206.055 g/mol
Opbevaringstemperatur: Fryser
MDL Number: MFCD11505926
CAS-nummer: 1027642-31-4

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR