4-Fluoro-2-(methylsulphonyl)toluene 95%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
4-Fluoro-1-methyl-2-(methylsulphonyl)benzene
PC904607-1G
PC904607-250MG
APOSPC904607-1GEA
5510
DKK
APOSPC904607-1G
APOSPC904607-250MG
4-Fluoro-2-(methylsulphonyl)toluene 95%
4-Fluoro-2-(methylsulphonyl)toluene
Formel:
C₈H₉FO₂S Molekylvægt: 188.219 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD09259030 CAS-nummer: 828270-66-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...