5-Fluoro-7-azaindole-3-carboxylic acid methyl ester 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PC904522-100MG PC904522-1G PC904522-250MG
APOSPC904522-100MGEA 1370 DKK
APOSPC904522-100MG APOSPC904522-1G APOSPC904522-250MG
5-Fluoro-7-azaindole-3-carboxylic acid methyl ester 95%
5-Fluoro-7-azaindole-3-carboxylic acid methyl ester
Formel: C₉H₇FN₂O₂
Molekylvægt: 194.162 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD12962794
CAS-nummer: 1027530-64-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR