6-Fluoro-2-methylbenzoxazole 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PC904514-1G PC904514-250MG
APOSPC904514-1GEA 3270 DKK
APOSPC904514-1G APOSPC904514-250MG
6-Fluoro-2-methylbenzoxazole 95%
6-Fluoro-2-methylbenzoxazole
Formel: C₈H₆FNO
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD03093954
CAS-nummer: 312600-96-7

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR