Ethyl 5-amino-2-fluorobenzoate 95%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
PC903640-1G
PC903640-5G
APOSPC903640-1GEA
162
DKK
APOSPC903640-1G
APOSPC903640-5G
Ethyl 5-amino-2-fluorobenzoate 95%
Ethyl 5-amino-2-fluorobenzoate
Formel:
C₉H₁₀FNO₂ Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD09037896 CAS-nummer: 123207-39-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...