1-[4-Bromo-3-(trifluoromethyl)phenyl]pyrrolidine 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

PC902128-1G PC902128-25G PC902128-5G
APOSPC902128-1GEA 850 DKK
APOSPC902128-1G APOSPC902128-25G APOSPC902128-5G
1-[4-Bromo-3-(trifluoromethyl)phenyl]pyrrolidine 98%
1-[4-Bromo-3-(trifluoromethyl)phenyl]pyrrolidine
Formel: C₁₁H₁₁BrF₃N
Molekylvægt: 294.111 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD09972140
CAS-nummer: 1020252-86-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR