5-Chloro-3-fluoro-2-hydroxypyridine 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PC900618-1G PC900618-5G
APOSPC900618-1GEA 194 DKK
APOSPC900618-1G APOSPC900618-5G
5-Chloro-3-fluoro-2-hydroxypyridine 98%
5-Chloro-3-fluoro-2-hydroxypyridine
Formel: C₅H₃ClFNO
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD07375044
CAS-nummer: 514797-96-7

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR