4-Chloro-2,3,6-trifluorobenzyl alcohol

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: (4-Chloro-2,3,6-trifluorophenyl)methanol

PC8258-1G PC8258-5G
APOSPC8258-1GEA 1490 DKK
APOSPC8258-1G APOSPC8258-5G
4-Chloro-2,3,6-trifluorobenzyl alcohol
4-Chloro-2,3,6-trifluorobenzyl alcohol
Formel: C₇H₄ClF₃O
Smeltepunkt: 38…43 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD03094475
CAS-nummer: 252004-68-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR