Sodium perfluorohexanoate 98%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
PC7314-25G
PC7314-5G
APOSPC7314-25GEA
9070
DKK
APOSPC7314-25G
APOSPC7314-5G
Sodium perfluorohexanoate 98%
Sodium perfluorohexanoate
Formel:
C₆F₁₁O₂·Na Smeltepunkt: 265…266 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD07784242 CAS-nummer: 2923-26-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...