4-Fluorobenzoic acid N-methylhydrazide

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 4-Fluoro-N-methylbenzohydrazide

PC6786-1G PC6786-250MG PC6786-5G
APOSPC6786-1GEA 502 DKK
APOSPC6786-1G APOSPC6786-250MG APOSPC6786-5G
4-Fluorobenzoic acid N-methylhydrazide
4-Fluorobenzoic acid N-methylhydrazide
Formel: C₈H₉FN₂O
Flammepunkt: >110 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD08461627
CAS-nummer: 94401-21-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR