(2S)-2-Fluoropropan-1-ol 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

PC6609-1G PC6609-250MG
APOSPC6609-1GEA 9330 DKK
APOSPC6609-1G APOSPC6609-250MG
(2S)-2-Fluoropropan-1-ol 97%
(2S)-2-Fluoropropan-1-ol
Formel: C₃H₇FO
Kogepunkt: 115 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD13176609
CAS-nummer: 877822-87-2
UN nummer: 1987
ADR: 3,III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR