Perfluoro-4-methoxybutanoic acid

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

PC6491-1G PC6491-5G PC6491-250MG
APOSPC6491-1GEA 1090 DKK
APOSPC6491-1G APOSPC6491-5G APOSPC6491-250MG
Perfluoro-4-methoxybutanoic acid
Perfluoro-4-methoxybutanoic acid
Formel: C₅HF₉O₃
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD07784238
CAS-nummer: 863090-89-5
UN nummer: 3265
ADR: 8,III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR