6-Bromo-3-chloro-2-fluorobenzaldehyde

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

PC57054-1G PC57054-250MG PC57054-5G
APOSPC57054-1GEA 349 DKK
APOSPC57054-1G APOSPC57054-250MG APOSPC57054-5G
6-Bromo-3-chloro-2-fluorobenzaldehyde
6-Bromo-3-chloro-2-fluorobenzaldehyde
Formel: C₇H₃BrClFO
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD11520661
CAS-nummer: 1114809-02-7

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR