2-Fluoro-3-methoxy-4-nitroaniline 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

PC56957-1G PC56957-250MG
APOSPC56957-1GEA 4320 DKK
APOSPC56957-1G APOSPC56957-250MG
2-Fluoro-3-methoxy-4-nitroaniline 95%
2-Fluoro-3-methoxy-4-nitroaniline
Formel: C₇H₇FN₂O₃
Molekylvægt: 186.14 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD31722920
CAS-nummer: 2091910-19-7

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR