Ethyl 2-(4-bromo-2-fluoro-6-formyl-phenoxy)acetate
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
PC56918-1G
PC56918-250MG
APOSPC56918-1GEA
4910
DKK
APOSPC56918-1G
APOSPC56918-250MG
Ethyl 2-(4-bromo-2-fluoro-6-formyl-phenoxy)acetate
Ethyl 2-(4-bromo-2-fluoro-6-formyl-phenoxy)acetate
Formel:
C₁₁H₁₀BrFO₄ Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD31695769 CAS-nummer: 2161554-52-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...