3-(Difluoromethyl)cyclobutanone 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 3-(Difluoromethyl)cyclobutan-1-one

PC56281-1G PC56281-250MG
APOSPC56281-1GEA 15340 DKK
APOSPC56281-1G APOSPC56281-250MG
3-(Difluoromethyl)cyclobutanone 95%
3-(Difluoromethyl)cyclobutanone
Formel: C₅H₆F₂O
Opbevaringstemperatur: Fryser
MDL Number: MFCD25371848
CAS-nummer: 1461713-56-3

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR