Tris(1H,1H,7H-perfluoroheptyl)borate
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
PC56217-1G
PC56217-250MG
APOSPC56217-1GEA
4360
DKK
APOSPC56217-1G
APOSPC56217-250MG
Tris(1H,1H,7H-perfluoroheptyl)borate
Tris(1H,1H,7H-perfluoroheptyl)borate
Formel:
C₂₁H₉BF₃₆O₃ Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD28137329 CAS-nummer: 1995-96-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...