2-Fluoro-3-iodobenzonitrile

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

PC53550-1G PC53550-5G
APOSPC53550-1GEA 1180 DKK
APOSPC53550-1G APOSPC53550-5G
2-Fluoro-3-iodobenzonitrile
2-Fluoro-3-iodobenzonitrile
Formel: C₇H₃FIN
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD18396938
CAS-nummer: 211943-27-0

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR