3-Fluoro-5-methoxycarbonylbenzeneboronic acid, pinacol ester

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PC53153-1G PC53153-5G
APOSPC53153-1GEA 1540 DKK
APOSPC53153-1G APOSPC53153-5G
3-Fluoro-5-methoxycarbonylbenzeneboronic acid, pinacol ester
3-Fluoro-5-methoxycarbonylbenzeneboronic acid, pinacol ester
Formel: C₁₄H₁₈BFO₄
Opbevaringstemperatur: Køleskab
MDL Number: MFCD12546616
CAS-nummer: 1016979-31-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR