6-Bromo-2,3-difluoronitrobenzene 95+%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 1-Bromo-3,4-difluoro-2-nitrobenzene

PC53041-1G PC53041-250MG
APOSPC53041-1GEA 1240 DKK
APOSPC53041-1G APOSPC53041-250MG
6-Bromo-2,3-difluoronitrobenzene 95+%
6-Bromo-2,3-difluoronitrobenzene
Formel: C₆H₂BrF₂NO₂
Molekylvægt: 237.986 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD03840797
CAS-nummer: 884495-47-0

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR