N-Benzyl-3-(trifluoromethoxy)-DL-phenylglycine 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PC52472-1G PC52472-250MG PC52472-5G
APOSPC52472-1GEA 3930 DKK
APOSPC52472-1G APOSPC52472-250MG APOSPC52472-5G
N-Benzyl-3-(trifluoromethoxy)-DL-phenylglycine 95%
N-Benzyl-3-(trifluoromethoxy)-DL-phenylglycine
Formel: C₁₆H₁₄F₃NO₃
Opbevaringstemperatur: Stue temperatur
CAS-nummer: 2514942-07-3

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR