3-Bromo-5-chloro-2-fluorobenzyl alcohol 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PC52453-1G PC52453-250MG PC52453-5G
APOSPC52453-1GEA 2080 DKK
APOSPC52453-1G APOSPC52453-250MG APOSPC52453-5G
3-Bromo-5-chloro-2-fluorobenzyl alcohol 95%
3-Bromo-5-chloro-2-fluorobenzyl alcohol
Formel: C₇H₅BrClFO
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD29036613
CAS-nummer: 1809161-65-6

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR