2-Bromo-5-iodobenzal fluoride
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
PC52189-1G
PC52189-250MG
PC52189-5G
APOSPC52189-1GEA
4320
DKK
APOSPC52189-1G
APOSPC52189-250MG
APOSPC52189-5G
2-Bromo-5-iodobenzal fluoride
2-Bromo-5-iodobenzal fluoride
Formel:
C₇H₄BrF₂I Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD18391113 CAS-nummer: 1261496-16-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...