5-(Trifluoromethyl)benzo[D]isoxazol-3-amine

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 5-(Trifluoromethyl)benzo[d]isoxazol-3-amine

PC52159-1G PC52159-250MG
APOSPC52159-1GEA 1530 DKK
APOSPC52159-1G APOSPC52159-250MG
5-(Trifluoromethyl)benzo[D]isoxazol-3-amine
5-(Trifluoromethyl)benzo[D]isoxazol-3-amine
Formel: C₈H₅F₃N₂O
Molekylvægt: 202.133 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD11847114
CAS-nummer: 868271-14-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR